siRNA Details
Gene Symbol: A330023F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015566.2 | 2 | 238 | |||
GenBank mRNA | AK039326.1 | - | 592 | |||
AK045637.1 | - | 2 | 239 | |||
AK171382.1 | - | 225 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB644138.1 | - | 2 | 239 | Chr. 1 - 195017399 - 195021564 [+] on Build GRCm38 |
