siRNA Details
Gene Symbol: 2900024J01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013590.1 | - | 1 | 211 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712550.1 | - | 1 | 202 | Chr. 1 - 180777850 - 180778762 [+] on Build GRCm38 |
