siRNA Details
Gene Symbol: C030015E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021085.1 | - | 1 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728168.1 | - | 1 | 304 | Chr. 5 - 48997851 - 48998536 [-] on Build GRCm38 |
