siRNA Details
Gene Symbol: C030014O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021082.1 | - | 1 | 513 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728165.1 | - | 1 | 510 | Chr. 2 - 130840195 - 130840871 [+] on Build GRCm38 |
