siRNA Details
Gene Symbol: 2210008F06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008669.1 | - | 2 | 682 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708573.1 | - | 2 | 682 | Chr. 11 - 77489854 - 77491292 [+] on Build GRCm38 |
