siRNA Details
Gene Symbol: 1700018H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006090.1 | - | 1 | 292 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV041509.2 | - | 1 | 143 | Chr. 11 - 83744591 - 83744867 [+] on Build GRCm38 |
