siRNA Details
Gene Symbol: 1700041I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006673.1 | - | 70 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706659.1 | - | 1 | 70 | Chr. 5 - 146795918 - 146806435 [+] on Build GRCm38 |
