siRNA Details
Gene Symbol: 1700041L08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006675.1 | - | 1 | 414 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DT911201.1 | - | 1 | 153 | Chr. 4 - 133672979 - 133673232 [+] on Build GRCm38 |
