siRNA Details
Gene Symbol: 1700065O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045386.1 | 5 | 687 | |||
GenBank mRNA | AK006898.1 | - | 5 | 690 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706891.1 | - | 5 | 689 | Chr. 18 - 49803326 - 49817958 [-] on Build GRCm38 |
