siRNA Details
Gene Symbol: Gm2848 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046069.1 | 1 | 76 | |||
GenBank mRNA | BC065398.1 | - | 1 | 76 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm2848 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046069.1 | 1 | 76 | |||
GenBank mRNA | BC065398.1 | - | 1 | 76 |