siRNA Details
Gene Symbol: Gm2848 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046069.1 | 4 | 414 | |||
GenBank mRNA | BC065398.1 | - | 4 | 414 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA118315.1 | - | 4 | 413 | Chr. 13 - 52570178 - 52573223 [-] on Build GRCm38 |
