siRNA Details
Gene Symbol: 2010100M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008298.1 | - | 1 | 32 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708231.1 | - | 1 | 29 | Chr. 19 - 41481265 - 41482182 [-] on Build GRCm38 |
