siRNA Details
Gene Symbol: 4930480K15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045463.1 | 3 | 2104 | |||
GenBank mRNA | AK019650.1 | - | 3 | 2103 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930480K15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045463.1 | 3 | 2104 | |||
GenBank mRNA | AK019650.1 | - | 3 | 2103 |