siRNA Details
Gene Symbol: 4930480K15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045463.1 | 1 | 550 | |||
GenBank mRNA | AK019650.1 | - | 1 | 551 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715153.1 | - | 1 | 550 | Chr. 17 - 90861169 - 90863023 [+] on Build GRCm38 |
