siRNA Details
Gene Symbol: A930007D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA575784.1 | - | 1 | 134 | ||
AK020824.1 | - | 1 | 21 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A930007D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA575784.1 | - | 1 | 134 | ||
AK020824.1 | - | 1 | 21 |