siRNA Details
Gene Symbol: 2310067P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010100.1 | - | 1 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709636.1 | - | 1 | 287 | Chr. 13 - 83065457 - 83194259 [+] on Build GRCm38 |
