siRNA Details
Gene Symbol: 2310068C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010104.1 | - | 1 | 1001 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DT924687.1 | - | 1 | 86 | Chr. 14 - 66991180 - 66991491 [-] on Build GRCm38 |
