siRNA Details
Gene Symbol: 1810013A23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045427.1 | 2 | 207 | |||
GenBank mRNA | AK007468.1 | - | 2 | 208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707535.1 | - | 2 | 208 | Chr. 17 - 28271708 - 28273163 [+] on Build GRCm38 |
