siRNA Details
Gene Symbol: Gm12273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052659.1 | - | 3 | 466 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660990.1 | - | 3 | 466 | Chr. 11 - 61950915 - 61952275 [+] on Build GRCm38 |
