siRNA Details
Gene Symbol: A730082K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040317.1 | 2, 3 | 382 | |||
NR_040319.1 | 2, 3 | 193 | ||||
GenBank mRNA | AK136021.1 | - | 2, 3 | 179 | ||
BG082928.2 | - | 2, 3 | 160 | |||
DV654286.1 | - | 2, 3 | 382 |
