siRNA Details
Gene Symbol: A730082K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040317.1 | 3 | 440 | |||
NR_040319.1 | 3 | 251 | ||||
GenBank mRNA | AK136021.1 | - | 3 | 237 | ||
BG082928.2 | - | 3 | 218 | |||
DV654286.1 | - | 3 | 440 |
