siRNA Details
Gene Symbol: A730082K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136021.1 | - | 1, 2 | 71 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ052895.1 | - | 1, 2 | 71 | Chr. 7 - 114951274 - 114975329 [+] on Build GRCm38 |
