siRNA Details
Gene Symbol: 2700080J24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012507.1 | - | 1 | 367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711632.1 | - | 1 | 367 | Chr. 7 - 16890088 - 16891067 [-] on Build GRCm38 |
