siRNA Details
Gene Symbol: 2700070H01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046019.1 | 3 | 510 | |||
GenBank mRNA | AK012509.1 | - | 3 | 510 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711634.1 | - | 3 | 513 | Chr. 14 - 63055073 - 63056940 [+] on Build GRCm38 |
