siRNA Details
Gene Symbol: 4930550I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016087.1 | - | 1 | 197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715689.1 | - | 1 | 197 | Chr. 2 - 72629018 - 72630432 [-] on Build GRCm38 |
