siRNA Details
Gene Symbol: Gm26629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039031.1 | - | 2 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632847.1 | - | 2 | 357 | Chr. 19 - 50677406 - 50744281 [+] on Build GRCm38 |
