siRNA Details
Gene Symbol: Lipt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039030.1 | - | 361 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632846.1 | - | 2 | 357 | Chr. 1 - 37865302 - 37868750 [+] on Build GRCm38 |
