siRNA Details
Gene Symbol: 2310021N16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009452.1 | - | 1 | 572 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709178.1 | - | 1 | 571 | Chr. 2 - 148280475 - 148281157 [-] on Build GRCm38 |
