siRNA Details
Gene Symbol: Gm12216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039036.1 | - | 3 | 245 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632853.1 | - | 3 | 245 | Chr. 11 - 53812948 - 53850077 [-] on Build GRCm38 |
