siRNA Details
Gene Symbol: 4632404H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035453.1 | - | 1 | 149 | ||
AK048101.1 | - | 1 | 3334 | |||
BC063049.1 | - | 1 | 2725 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627495.1 | - | 1 | 149 | Chr. 3 - 89769598 - 89770215 [-] on Build GRCm38 |
