siRNA Details
Gene Symbol: 2810461L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013369.1 | - | 1 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712363.1 | - | 1 | 78 | Chr. 2 - 180890684 - 180890968 [+] on Build GRCm38 |
