siRNA Details
Gene Symbol: Uba6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017687.1 | - | 788 | |||
AK029740.1 | - | 2072 | ||||
AK139327.1 | - | 1230 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ257842.1 | - | 1 | 547 | Chr. 5 - 86109507 - 86110201 [-] on Build GRCm38 |
