siRNA Details
Gene Symbol: 0610042B23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002901.1 | - | 1 | 79 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703398.1 | - | 1 | 77 | Chr. 8 - 126946088 - 126946477 [+] on Build GRCm38 |
