siRNA Details
Gene Symbol: Gm30223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083219.1 | - | 2 | 536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB653349.1 | - | 2 | 536 | Chr. 3 - 16817092 - 16824180 [+] on Build GRCm38 |
