siRNA Details
Gene Symbol: 2900084I15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013819.1 | - | 1 | 372 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712784.1 | - | 1 | 340 | Chr. 6 - 126138690 - 126139191 [-] on Build GRCm38 |
