siRNA Details
Gene Symbol: Gm26560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044390.1 | - | 3 | 2448 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB278360.2 | - | 3 | 504 | Chr. 5 - 38317336 - 38318942 [-] on Build GRCm38 |
