siRNA Details
Gene Symbol: Gm31367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133333.1 | - | 200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB616744.1 | - | 2 | 200 | Chr. 8 - 116605951 - 116622799 [+] on Build GRCm38 |
