siRNA Details
Gene Symbol: 6720451E02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020122.1 | - | 1 | 81 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719340.1 | - | 1 | 78 | Chr. 3 - 55833810 - 55834458 [-] on Build GRCm38 |
