siRNA Details
Gene Symbol: 6720454L07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020125.1 | - | 1 | 67 | ||
AK137081.1 | - | 1 | 877 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719350.1 | - | 1 | 67 | Chr. 11 - 88532048 - 88532692 [-] on Build GRCm38 |
