siRNA Details
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003552.1 | - | 1 | 522 | ||
AK151082.1 | - | 1039 | ||||
AK153482.1 | - | 1040 | ||||
AK184946.1 | - | 1 | 396 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003552.1 | - | 1 | 522 | ||
AK151082.1 | - | 1039 | ||||
AK153482.1 | - | 1040 | ||||
AK184946.1 | - | 1 | 396 |