siRNA Details
Gene Symbol: Gm20513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153671.1 | - | 2 | 164 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ137617.1 | - | 2 | 164 | Chr. 17 - 34293071 - 34305720 [-] on Build GRCm38 |
