siRNA Details
Gene Symbol: 8430426J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077229.1 | 2 | 959 | |||
NR_077230.1 | 2 | 826 | ||||
NR_077231.1 | 2 | 720 | ||||
GenBank mRNA | AV290832.1 | - | 1 | 188 | ||
BC038155.1 | - | 2 | 826 | |||
BC051511.1 | - | 2 | 725 | |||
BC062269.1 | - | 1 | 563 |
