siRNA Details
Gene Symbol: 2310076G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039736.1 | - | 1 | 1365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX240119.1 | - | 1 | 112 | Chr. 9 - 81628764 - 81629546 [-] on Build GRCm38 |
