siRNA Details
Gene Symbol: 4930533P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040478.1 | 3 | 350 | |||
GenBank mRNA | AK015960.1 | - | 3 | 351 | ||
BY715526.1 | - | 3 | 348 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930533P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040478.1 | 3 | 350 | |||
GenBank mRNA | AK015960.1 | - | 3 | 351 | ||
BY715526.1 | - | 3 | 348 |