siRNA Details
Gene Symbol: A330069E16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132032.1 | - | 320 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ044612.1 | - | 1 | 320 | Chr. 2 - 91236487 - 91237447 [+] on Build GRCm38 |
