siRNA Details
Gene Symbol: Gm16998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038016.1 | 2 | 956 | |||
GenBank mRNA | AK138755.1 | - | 2 | 957 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB202760.1 | - | 1 | 41 | Chr. 10 - 54078220 - 54081074 [+] on Build GRCm38 |
