siRNA Details
Gene Symbol: Gm16998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038016.1 | 2, 3 | 1105 | |||
GenBank mRNA | AK138755.1 | - | 2, 3 | 1106 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038016.1 | 2, 3 | 1105 | |||
GenBank mRNA | AK138755.1 | - | 2, 3 | 1106 |