siRNA Details
Gene Symbol: 9330164J24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034219.1 | - | 1 | 1506 | ||
AK039693.1 | - | 1 | 1735 | |||
AK086125.1 | - | 1 | 1484 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB080741.2 | - | 1 | 103 | Chr. 17 - 90800997 - 90801656 [-] on Build GRCm38 |
