siRNA Details
Gene Symbol: 9330164J24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034219.1 | - | 1 | 1928 | ||
AK039693.1 | - | 1 | 2157 | |||
AK086125.1 | - | 1 | 1906 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9330164J24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034219.1 | - | 1 | 1928 | ||
AK039693.1 | - | 1 | 2157 | |||
AK086125.1 | - | 1 | 1906 |