siRNA Details
Gene Symbol: Gm34517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132743.1 | - | 1 | 45 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ046802.1 | - | 1 | 45 | Chr. 19 - 43266936 - 43277499 [+] on Build GRCm38 |
